Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,6-Dimethyl-3-oxabicyclo[3.1.0]hexane-2,4-dione, 98%, Alfa Aesar™

Click to view available options
Quantity:
25g
100g
67911-21-1
C7H8O3
140.138
MFCD09751198
QKAHKEDLPBJLFD-UHFFFAOYSA-N
6,6-dimethyl-3-oxabicyclo 3.1.0 hexane-2,4-dione,caronic anhydride,3-oxabicyclo 3.1.0 hexane-2,4-dione, 6,6-dimethyl,3,3-dimethyl-1,2-cyclopropanedicarboxylic anhydride,ksc626s3r,6,6-dimethyl-3-oxabicyclo 3.1.0 hex-2,4-dione,3,3-dimethyl-1,2-cyclopropane dicarboxylic anhydride,3-oxabicyclo 3.1.0 hexane-2,4-dione,6,6-dimethyl,3,3-dimethyl-1,2-cyclopropanedicarboxylic acid anhydride,2r,4r-methyl 4-hydroxypyrrolidine-2-carboxylate hydrochloride form
10374536
6,6-dimethyl-3-oxabicyclo[3.1.0]hexane-2,4-dione
CC1(C2C1C(=O)OC2=O)C
67911-21-1
C7H8O3
140.138
MFCD09751198
QKAHKEDLPBJLFD-UHFFFAOYSA-N
6,6-dimethyl-3-oxabicyclo 3.1.0 hexane-2,4-dione,caronic anhydride,3-oxabicyclo 3.1.0 hexane-2,4-dione, 6,6-dimethyl,3,3-dimethyl-1,2-cyclopropanedicarboxylic anhydride,ksc626s3r,6,6-dimethyl-3-oxabicyclo 3.1.0 hex-2,4-dione,3,3-dimethyl-1,2-cyclopropane dicarboxylic anhydride,3-oxabicyclo 3.1.0 hexane-2,4-dione,6,6-dimethyl,3,3-dimethyl-1,2-cyclopropanedicarboxylic acid anhydride,2r,4r-methyl 4-hydroxypyrrolidine-2-carboxylate hydrochloride form
10374536
6,6-dimethyl-3-oxabicyclo[3.1.0]hexane-2,4-dione
CC1(C2C1C(=O)OC2=O)C
Color White
Melting Point 53°C to 57°C
Boiling Point 137°C (14mmHg)
Quantity 100g
Sensitivity Moisture Sensitive
Beilstein 117391
Formula Weight 140.14
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 6,6-Dimethyl-3-oxabicyclo[3.1.0]hexane-2,4-dione

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning