Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,7-Dihydro-5H-pyrrolo[3,4-d]pyrimidine dihydrochloride, 95%, Thermo Scientific™

Catalog Number ac454212500
Click to view available options
Quantity:
250 mg
1 g
157327-51-0
C6H7N3·2ClH
194.06
VGQSNQPVXPPLEH-UHFFFAOYSA-N
6,7-dihydro-5h-pyrrolo 3,4-d pyrimidine dihydrochloride, 5h,6h,7h-pyrrolo 3,4-d pyrimidine dihydrochloride, 6,7-dihydro-5h-pyrrolo 3,4-d pyrimidine 2hcl, 6,7-dihydro-5h-pyrrolo-3,4-d-pyrimidine dihydrochloride, 4-d pyrimidine hydrochloride, 6,7-dihydro-5h-pyrrolo 3,4-d pyrimidinedihydrochloride, 6,7-dihydro-5h-pyrrolo 3,4-d pyrimidine dihydrochl, 5h-pyrrolo 3,4-d pyrimidine,6,7-dihydro-, hydrochloride, 5h-pyrrolo 3,4-d pyrimidine,6,7-dihydro-,dihydrochloride, 5h-pyrrolo 3,4-d pyrimidine, 6,7-dihydro-, hydrochloride 1:2
44182365
6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidine;dihydrochloride
C1C2=CN=CN=C2CN1.Cl.Cl
157327-51-0
C6H7N3·2ClH
194.06
VGQSNQPVXPPLEH-UHFFFAOYSA-N
6,7-dihydro-5h-pyrrolo 3,4-d pyrimidine dihydrochloride, 5h,6h,7h-pyrrolo 3,4-d pyrimidine dihydrochloride, 6,7-dihydro-5h-pyrrolo 3,4-d pyrimidine 2hcl, 6,7-dihydro-5h-pyrrolo-3,4-d-pyrimidine dihydrochloride, 4-d pyrimidine hydrochloride, 6,7-dihydro-5h-pyrrolo 3,4-d pyrimidinedihydrochloride, 6,7-dihydro-5h-pyrrolo 3,4-d pyrimidine dihydrochl, 5h-pyrrolo 3,4-d pyrimidine,6,7-dihydro-, hydrochloride, 5h-pyrrolo 3,4-d pyrimidine,6,7-dihydro-,dihydrochloride, 5h-pyrrolo 3,4-d pyrimidine, 6,7-dihydro-, hydrochloride 1:2
44182365
6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidine;dihydrochloride
C1C2=CN=CN=C2CN1.Cl.Cl
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Argentometry)
Quantity 250 mg
Formula Weight 194.06
Percent Purity 95%
Chemical Name or Material 6,7-Dihydro-5H-pyrrolo[3,4-d]pyrimidine dihydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit