Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, 97%

Click to view available options
Quantity:
5g
25g
100g
2328-12-3
C11H16ClNO2
229.70
MFCD00012744
SHOWAGCIRTUYNA-UHFFFAOYSA-N
6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride,heliamine hydrochloride,6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolinehydrochloride,6,7-dimethoxy-1,2,3,4-tetrahydro-isoquinoline hydrochloride,1,2,3,4-tetrahydro-6,7-dimethoxy-isoquinoline hydrochloride,isoquinoline, 1,2,3,4-tetrahydro-6,7-dimethoxy-, hydrochloride,6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline, chloride,pubchem22149,acmc-1cjku,ksc203s7f
6456035
6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline;hydrochloride
COC1=C(C=C2CNCCC2=C1)OC.Cl
2328-12-3
C11H16ClNO2
229.70
MFCD00012744
SHOWAGCIRTUYNA-UHFFFAOYSA-N
6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride,heliamine hydrochloride,6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolinehydrochloride,6,7-dimethoxy-1,2,3,4-tetrahydro-isoquinoline hydrochloride,1,2,3,4-tetrahydro-6,7-dimethoxy-isoquinoline hydrochloride,isoquinoline, 1,2,3,4-tetrahydro-6,7-dimethoxy-, hydrochloride,6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline, chloride,pubchem22149,acmc-1cjku,ksc203s7f
6456035
6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline;hydrochloride
COC1=C(C=C2CNCCC2=C1)OC.Cl
Melting Point 256°C to 260°C
Quantity 5g
Beilstein 3634126
Formula Weight 229.71
Percent Purity 97%
Chemical Name or Material 6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : NX5017980