Alfa Aesar™ 6-Acetamidohexanoic acid hydrate, 99%

Manufacturer: Alfa Aesar™ B2362314

Quantity 25g

 View more versions of this product

Catalog No. AAB2362314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 57-08-9
    Molecular Formula C8H15NO3·xH2O
    Linear Formula CH3CONH(CH2)5CO2H·xH2O
    Formula Weight 173.21 (Anhydrous)
    MDL Number MFCD00004424
    Merck Index 14,45
    Synonym 6-Acetamidocaproic acid hydrate
    Chemical Name or Material 6-Acetamidohexanoic acid hydrate

    Specifications

    Formula Weight 173.21 (Anhydrous)
    Molecular Formula C8H15NO3·xH2O
    Linear Formula CH3CONH(CH2)5CO2H·xH2O
    MDL Number MFCD00004424
    CAS 57-08-9
    Merck Index 14,45
    Synonym 6-Acetamidocaproic acid hydrate
    Melting Point 102° to 106°C
    Quantity 25g
    Chemical Name or Material 6-Acetamidohexanoic acid hydrate
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit