Alfa Aesar™ 6-Amino-1-methyluracil, 97%

Manufacturer: Alfa Aesar™ B2544822

Quantity 100g

 View more versions of this product

Catalog No. AAB2544822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2434-53-9
    Molecular Formula C5H7N3O2
    Formula Weight 141.13
    MDL Number MFCD00075366
    Beilstein 127771
    Synonym 6-Amino-1-methyl-2,4(1H,3H)-pyrimidinedione
    Chemical Name or Material 6-Amino-1-methyluracil

    Specifications

    Formula Weight 141.13
    MDL Number MFCD00075366
    Melting Point >300°C
    Quantity 100g
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Molecular Formula C5H7N3O2
    CAS 2434-53-9
    Beilstein 127771
    Synonym 6-Amino-1-methyl-2,4(1H,3H)-pyrimidinedione
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 6-Amino-1-methyluracil
    Assay 97%

    Description

    Applications
    6-Amino-1-methyluracil may be used in the preparation of 1,1 ft.-di methyl-1H-spiro[pyrimido[4,5-b]quinoline-5,5 ft.-pyrrolo[2,3-d]pyrimidine]-2,2 ft.,4,4 ft.,6 ft.(1 ft.H,3H,3 ft.H,7 ft.H,1 ft.H)-pentaone, via reaction with isatin in the presence of catalytic p-toluene sulfonic acid.

    Solubility
    Soluble in diluted sodium hydroxide solution.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit