Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Amino-1-methyluracil, 97%

Click to view available options
Quantity:
25g
100g
2434-53-9
C5H7N3O2
141.13
MFCD00075366
GZLZRPNUDBIQBM-UHFFFAOYSA-N
6-amino-1-methyluracil,6-amino-1-methylpyrimidine-2,4 1h,3h-dione,1-methyl-6-aminouracil,2,4 1h,3h-pyrimidinedione, 6-amino-1-methyl,6-amino-1-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione,6-amino-1-methyl-2,4 1h,3h-pyrimidinedione,6-amino-1-methyl-1h-pyrimidine-2,4-dione,6-amino-1-methyl-1,3-dihydropyrimidine-2,4-dione,6-amino-1-methyl-3h-pyrimidine-2,4-dione,zlchem 437
75520
6-amino-1-methylpyrimidine-2,4-dione
CN1C(=CC(=O)NC1=O)N
2434-53-9
C5H7N3O2
141.13
MFCD00075366
GZLZRPNUDBIQBM-UHFFFAOYSA-N
6-amino-1-methyluracil,6-amino-1-methylpyrimidine-2,4 1h,3h-dione,1-methyl-6-aminouracil,2,4 1h,3h-pyrimidinedione, 6-amino-1-methyl,6-amino-1-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione,6-amino-1-methyl-2,4 1h,3h-pyrimidinedione,6-amino-1-methyl-1h-pyrimidine-2,4-dione,6-amino-1-methyl-1,3-dihydropyrimidine-2,4-dione,6-amino-1-methyl-3h-pyrimidine-2,4-dione,zlchem 437
75520
6-amino-1-methylpyrimidine-2,4-dione
CN1C(=CC(=O)NC1=O)N
Applications
6-Amino-1-methyluracil may be used in the preparation of 1,1′-di methyl-1H-spiro[pyrimido[4,5-b]quinoline-5,5′-pyrrolo[2,3-d]pyrimidine]-2,2′,4,4′,6′(1′H,3H,3′H,7′H,1′H)-pentaone, via reaction with isatin in the presence of catalytic p-toluene sulfonic acid.

Solubility
Soluble in diluted sodium hydroxide solution.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Beilstein 127771
Formula Weight 141.13
Percent Purity 97%
Melting Point >300°C
Quantity 25g
Odor Odorless
Chemical Name or Material 6-Amino-1-methyluracil

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning