Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Amino-1H-indazole, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
6967-12-0
C7H7N3
133.154
MFCD00005696
KEJFADGISRFLFO-UHFFFAOYSA-N
6-aminoindazole,6-amino-1h-indazole,2h-indazol-6-amine,1h-indazol-6-ylamine,6-amino-indazole,indazol-6-ylamine,6-aminoimidazole,6-aminobenzopyrazole,1h-indazole-6-ylamine,indazol-6-amine
81423
1H-indazol-6-amine
C1=CC2=C(C=C1N)NN=C2
6967-12-0
C7H7N3
133.154
MFCD00005696
KEJFADGISRFLFO-UHFFFAOYSA-N
6-aminoindazole,6-amino-1h-indazole,2h-indazol-6-amine,1h-indazol-6-ylamine,6-amino-indazole,indazol-6-ylamine,6-aminoimidazole,6-aminobenzopyrazole,1h-indazole-6-ylamine,indazol-6-amine
81423
1H-indazol-6-amine
C1=CC2=C(C=C1N)NN=C2
Infrared Spectrum Authentic
Melting Point 204°C to 210°C
Color Tan
Quantity 5g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 133.15
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 6-Amino-1H-indazole
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.