Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Amino-2,4-xylenol 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
41458-65-5
C8H11NO
137.182
MFCD00007691
GISWNAMJAQRJPC-UHFFFAOYSA-N
6-Amino-2,4-dimethylphenol, 2-Hydroxy-3,5-dimethylaniline
100530
2-amino-4,6-dimethylphenol
CC1=CC(=C(C(=C1)N)O)C
41458-65-5
C8H11NO
137.182
MFCD00007691
GISWNAMJAQRJPC-UHFFFAOYSA-N
6-Amino-2,4-dimethylphenol, 2-Hydroxy-3,5-dimethylaniline
100530
2-amino-4,6-dimethylphenol
CC1=CC(=C(C(=C1)N)O)C
Color Undesignated
Quantity 5g
Melting Point 130°C
Formula Weight 137.18
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 6-Amino-2,4-xylenol