Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6′-Amino-3′,4′-(methylenedioxy)acetophenone 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
28657-75-2
C9H9NO3
179.175
MFCD00075855
DWTHYSZSRJOMSC-UHFFFAOYSA-N
1-6-aminobenzo d 1,3 dioxol-5-yl ethanone,6'-amino-3',4'-methylenedioxy acetophenone,1-6-amino-1,3-benzodioxol-5-yl ethanone,2'-amino-4',5'-methylenedioxy acetophenone,2-amino-4,5-methylenedioxyacetophenone,1-6-amino-2h-1,3-benzodioxol-5-yl ethan-1-one,1-6-amino-1,3-benzodioxol-5-yl ethan-1-one,2'-amino-4',5'-methylenedioxyacetophenone,5-acetyl-6-amino-1,3-benzodioxole,1-6-amino-2h-1,3-benzodioxol-5-yl ethanone
120000
1-(6-amino-1,3-benzodioxol-5-yl)ethanone
CC(=O)C1=CC2=C(C=C1N)OCO2
28657-75-2
C9H9NO3
179.175
MFCD00075855
DWTHYSZSRJOMSC-UHFFFAOYSA-N
1-6-aminobenzo d 1,3 dioxol-5-yl ethanone,6'-amino-3',4'-methylenedioxy acetophenone,1-6-amino-1,3-benzodioxol-5-yl ethanone,2'-amino-4',5'-methylenedioxy acetophenone,2-amino-4,5-methylenedioxyacetophenone,1-6-amino-2h-1,3-benzodioxol-5-yl ethan-1-one,1-6-amino-1,3-benzodioxol-5-yl ethan-1-one,2'-amino-4',5'-methylenedioxyacetophenone,5-acetyl-6-amino-1,3-benzodioxole,1-6-amino-2h-1,3-benzodioxol-5-yl ethanone
120000
1-(6-amino-1,3-benzodioxol-5-yl)ethanone
CC(=O)C1=CC2=C(C=C1N)OCO2
Quantity 25g
Color Undesignated
Melting Point 171°C
Formula Weight 179.18
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 6′-Amino-3′,4′-(methylenedioxy)acetophenone