Learn More
Alfa Aesar™ 6-Amino-3-iodo-2-methylpyridine, 95%
Manufacturer:  Alfa Aesar™ H27265MD
Quantity 250mg
 View more versions of this product
Catalog No. AAH27265MD
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
75073-11-9
234.04
6-Amino-3-iodo-2-picoline; 5-Iodo-6-methyl-2-pyridinamine
C6H7IN2
MFCD04138418
6-Amino-3-iodo-2-methylpyridine
Specifications
75073-11-9
234.04
250mg
6-Amino-3-iodo-2-methylpyridine
C6H7IN2
MFCD04138418
6-Amino-3-iodo-2-picoline; 5-Iodo-6-methyl-2-pyridinamine
95%
Safety and Handling
SDS & Safety Documents
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger