Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Amino-7-deazapurine, 97%

Click to view available options
Quantity:
1g
5g
1500-85-2
C6H6N4
134.142
MFCD01686848
PEHVGBZKEYRQSX-UHFFFAOYSA-N
4-amino-7h-pyrrolo 2,3-d pyrimidine,7h-pyrrolo 2,3-d pyrimidin-4-amine,7-deazaadenine,7h-pyrrolo 2,3-d pyrimidin-4-ylamine,4-aminopyrrolo 2,3-d pyrimidine,4-aminopyrrolo 2,3-d pyrimidine,1h-pyrrolo 2,3-d pyrimidin-4-amine,7h-pyrrolo 2,3-d pyrimidine, 4-amino,7h-pyrrolo 2,3-d pyrimidine, 4-amino,pubchem14980
5359620
7H-pyrrolo[2,3-d]pyrimidin-4-amine
C1=CNC2=C1C(=NC=N2)N
1500-85-2
C6H6N4
134.142
MFCD01686848
PEHVGBZKEYRQSX-UHFFFAOYSA-N
4-amino-7h-pyrrolo 2,3-d pyrimidine,7h-pyrrolo 2,3-d pyrimidin-4-amine,7-deazaadenine,7h-pyrrolo 2,3-d pyrimidin-4-ylamine,4-aminopyrrolo 2,3-d pyrimidine,4-aminopyrrolo 2,3-d pyrimidine,1h-pyrrolo 2,3-d pyrimidin-4-amine,7h-pyrrolo 2,3-d pyrimidine, 4-amino,7h-pyrrolo 2,3-d pyrimidine, 4-amino,pubchem14980
5359620
7H-pyrrolo[2,3-d]pyrimidin-4-amine
C1=CNC2=C1C(=NC=N2)N
Sensitivity Moisture & Light Sensitive
Melting Point 257°C to 262°C
Color Brown to White
Quantity 5g
Formula Weight 134.14
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 6-Amino-7-deazapurine

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UY8850000

storageNote1 : 2° to 8°C