Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Amino-7-deazapurine hydrogen sulfate, 97%

Click to view available options
Quantity:
250mg
1g
5g
769951-32-8
C6H8N4O4S
232.214
MFCD09787504
ZVFLCRGXDSPVKK-UHFFFAOYSA-N
4-amino-7h-pyrrolo 2,3-d pyrimidine hydrogen sulfate,7h-pyrrolo 2,3-d pyrimidin-4-amine sulfate,7h-pyrrolo 2,3-d pyrimidin-4-amine sulphate,4-amino-7h-pyrrolo 2,3-d pyrimidine sulfate,7h-pyrrolo 2,3-d pyrimidin-4-amine; sulfuric acid,7-deazaadenine; sulfuric acid,1h-pyrrolo 2,3-d pyrimidin-4-amine sulfate,7h-pyrrolo 2,3-d pyrimidin-4-amine sulfuric acid,4-amino-7h-pyrrolo 2,3-d pyrimidinehydrogensulfate,ksc376q6n
11470294
7H-pyrrolo[2,3-d]pyrimidin-4-amine;sulfuric acid
C1=CNC2=C1C(=NC=N2)N.OS(=O)(=O)O
769951-32-8
C6H8N4O4S
232.214
MFCD09787504
ZVFLCRGXDSPVKK-UHFFFAOYSA-N
4-amino-7h-pyrrolo 2,3-d pyrimidine hydrogen sulfate,7h-pyrrolo 2,3-d pyrimidin-4-amine sulfate,7h-pyrrolo 2,3-d pyrimidin-4-amine sulphate,4-amino-7h-pyrrolo 2,3-d pyrimidine sulfate,7h-pyrrolo 2,3-d pyrimidin-4-amine; sulfuric acid,7-deazaadenine; sulfuric acid,1h-pyrrolo 2,3-d pyrimidin-4-amine sulfate,7h-pyrrolo 2,3-d pyrimidin-4-amine sulfuric acid,4-amino-7h-pyrrolo 2,3-d pyrimidinehydrogensulfate,ksc376q6n
11470294
7H-pyrrolo[2,3-d]pyrimidin-4-amine;sulfuric acid
C1=CNC2=C1C(=NC=N2)N.OS(=O)(=O)O
Color Brown
Quantity 1g
Formula Weight 232.21
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 6-Amino-7-deazapurine hydrogen sulfate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C