Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Amino-m-cresol 98.0+%, TCI America™

Click to view available options
Quantity:
25g
250g
2835-98-5
C7H9NO
123.155
MFCD00007693
HCPJEHJGFKWRFM-UHFFFAOYSA-N
6-amino-m-cresol,2-hydroxy-4-methylaniline,phenol, 2-amino-5-methyl,4-amino-3-hydroxytoluene,2-hydroxy-p-toluidine,unii-qcg4es2a26,6-amino-3-cresol,3-methyl-6-aminophenol,6-amino-3-methylphenol,qcg4es2a26
76082
2-amino-5-methylphenol
CC1=CC(=C(C=C1)N)O
2835-98-5
C7H9NO
123.155
MFCD00007693
HCPJEHJGFKWRFM-UHFFFAOYSA-N
6-amino-m-cresol,2-hydroxy-4-methylaniline,phenol, 2-amino-5-methyl,4-amino-3-hydroxytoluene,2-hydroxy-p-toluidine,unii-qcg4es2a26,6-amino-3-cresol,3-methyl-6-aminophenol,6-amino-3-methylphenol,qcg4es2a26
76082
2-amino-5-methylphenol
CC1=CC(=C(C=C1)N)O
Color Undesignated
Quantity 250g
Melting Point 159°C
Formula Weight 123.16
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 6-Amino-m-cresol

EINECSNumber : (3)-0707

TSCA : No