Alfa Aesar™ 6-Aminobenzoxazole, 95%

Manufacturer: Alfa Aesar™ H34485MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH34485MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 177492-52-3
    Formula Weight 134.1
    MDL Number MFCD03791187
    Synonym Benzoxazol-6-amine
    Chemical Name or Material 6-Aminobenzoxazole

    Specifications

    Melting Point 95° to 97°C
    Formula Weight 134.1
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD03791187
    CAS 177492-52-3
    Sensitivity Air Sensitive
    Health Hazard 2 GHS H Statement
    H301-H332-H315-H319-H335
    Toxic if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Physical Form Solid
    Color Orange to brown
    Quantity 250mg
    Health Hazard 1 Danger
    Storage Note 1 2° to 8°C
    Synonym Benzoxazol-6-amine
    Chemical Name or Material 6-Aminobenzoxazole
    Assay 95%

    Description

    Applications
    6-Aminobenzoxazole is used as pharmaceutical intermediate.

    Solubility
    Slightly soluble in water.

    Notes
    Air sensitive. Incompatible with oxidizing agents and heat.

    GHS H Statement
    H301-H332-H315-H319-H335
    Toxic if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    2° to 8°C

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit