Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Aminohexanenitrile, 98%

Manufacturer:  Alfa Aesar™ H2780406

Quantity 5g

 View more versions of this product

Catalog No. AAH2780406

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
Used mainly as intermediates. The results obtained by varying either the external reaction parameters or the internal catalyst parameters show 6-amino- hexanenitrile, 1,6-diaminohexane and azacycloheptane as the only products of hydrogenation of hexanedinitrile.

Solubility
Moderately soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

2432-74-8
112.17
6-Aminocapronitrile
H2N(CH2)5CN
MFCD00009822
6-Aminohexanenitrile

Specifications

2432-74-8
112.17
5g
6-Aminohexanenitrile
H2N(CH2)5CN
MFCD00009822
6-Aminocapronitrile
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : MO4085000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit