Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Aminoindan-1-one, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
69975-65-1
C9H9NO
147.177
MFCD00100747
UOJCPAAEKXNPQT-UHFFFAOYSA-N
6-aminoindanone,6-aminoindan-1-one,6-amino-1-indanone,6-amino-2,3-dihydro-1h-inden-1-one,6-amino-indan-1-one,1h-inden-1-one, 6-amino-2,3-dihydro,6-amino indanone,ksc497g1b,6-azanyl-2,3-dihydroinden-1-one
312895
6-amino-2,3-dihydroinden-1-one
C1CC(=O)C2=C1C=CC(=C2)N
69975-65-1
C9H9NO
147.177
MFCD00100747
UOJCPAAEKXNPQT-UHFFFAOYSA-N
6-aminoindanone,6-aminoindan-1-one,6-amino-1-indanone,6-amino-2,3-dihydro-1h-inden-1-one,6-amino-indan-1-one,1h-inden-1-one, 6-amino-2,3-dihydro,6-amino indanone,ksc497g1b,6-azanyl-2,3-dihydroinden-1-one
312895
6-amino-2,3-dihydroinden-1-one
C1CC(=O)C2=C1C=CC(=C2)N
Infrared Spectrum Authentic
Quantity 1g
Melting Point 156.0°C to 158.0°C
Color Black to Brown
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 147.18
Physical Form Solution
Percent Purity 97%
Chemical Name or Material 6-Aminoindan-1-one
Show More Show Less

GHS P Statement
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning