Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Aminoindole, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
5318-27-4
C8H8N2
132.17
MFCD02093959
MIMYTSWNVBMNRH-UHFFFAOYSA-N
6-aminoindole,6-amino-1h-indole,6-amino indole,1h-indol-6-ylamine,indole-6-ylamine,1h-indol-6-amin,6-indolamine,indol-6-amine,pubchem7311
256096
1H-indol-6-amine
C1=CC(=CC2=C1C=CN2)N
5318-27-4
C8H8N2
132.17
MFCD02093959
MIMYTSWNVBMNRH-UHFFFAOYSA-N
6-aminoindole,6-amino-1h-indole,6-amino indole,1h-indol-6-ylamine,indole-6-ylamine,1h-indol-6-amin,6-indolamine,indol-6-amine,pubchem7311
256096
1H-indol-6-amine
C1=CC(=CC2=C1C=CN2)N
Boiling Point 161.0°C to 166.0°C (2.3 Torr)
Infrared Spectrum Authentic
Melting Point 66.0°C to 68.0°C
Packaging Glass bottle
Quantity 5g
Color White to Brown
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 132.17
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 6-Aminoindole
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning