Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Aminoindole 98.0+%, TCI America™

Click to view available options
Quantity:
1g
5g
5318-27-4
C8H8N2
132.166
MFCD02093959
MIMYTSWNVBMNRH-UHFFFAOYSA-N
6-aminoindole,6-amino-1h-indole,6-amino indole,1h-indol-6-ylamine,indole-6-ylamine,1h-indol-6-amin,6-indolamine,indol-6-amine,pubchem7311
256096
1H-indol-6-amine
C1=CC(=CC2=C1C=CN2)N
5318-27-4
C8H8N2
132.166
MFCD02093959
MIMYTSWNVBMNRH-UHFFFAOYSA-N
6-aminoindole,6-amino-1h-indole,6-amino indole,1h-indol-6-ylamine,indole-6-ylamine,1h-indol-6-amin,6-indolamine,indol-6-amine,pubchem7311
256096
1H-indol-6-amine
C1=CC(=CC2=C1C=CN2)N
Quantity 5g
Boiling Point 166°C
Melting Point 72°C
Color Undesignated
Formula Weight 132.17
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 6-Aminoindole

TSCA : No

Recommended Storage : Refrigerator