Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Aminonicotinic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
3167-49-5
C6H6N2O2
138.13
MFCD00006326
ZCIFWRHIEBXBOY-UHFFFAOYSA-N
6-aminonicotinic acid, 3-pyridinecarboxylic acid, 6-amino, 2-amino-5-pyridinecarboxylic acid, 6-amino-nicotinic acid, 6-amino nicotinic acid, 2-aminopyridine-5-carboxylic acid, nicotinic acid, 6-amino, unii-9vr2o1ok60, 6-amino-3-pyridinecarboxylic acid, 6-aminopyridine 3-carboxylic acid
18496
CHEBI:68583
6-aminopyridine-3-carboxylic acid
C1=CC(=NC=C1C(=O)O)N
3167-49-5
C6H6N2O2
138.13
MFCD00006326
ZCIFWRHIEBXBOY-UHFFFAOYSA-N
6-aminonicotinic acid, 3-pyridinecarboxylic acid, 6-amino, 2-amino-5-pyridinecarboxylic acid, 6-amino-nicotinic acid, 6-amino nicotinic acid, 2-aminopyridine-5-carboxylic acid, nicotinic acid, 6-amino, unii-9vr2o1ok60, 6-amino-3-pyridinecarboxylic acid, 6-aminopyridine 3-carboxylic acid
18496
CHEBI:68583
6-aminopyridine-3-carboxylic acid
C1=CC(=NC=C1C(=O)O)N
Melting Point >300°C
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
Color White to Beige
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula H2N(C5H3N)COOH
Beilstein 22, 542
Merck Index 15, 452
Solubility Information Solubility in water: ca 1.0g/L (20°C). Other solubilities: sparingly soluble in most solvents,5.0g/L (20°C) in ethanol
Formula Weight 138.13
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 6-Aminonicotinic acid, 98%
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

EINECSNumber : 221-630-4

TSCA : TSCA