Alfa Aesar™ 6-Aminopenicillanic acid, 98%

Manufacturer: Alfa Aesar™ A1305609

Quantity 10g

 View more versions of this product

Catalog No. AAA1305609

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 551-16-6
    Molecular Formula C8H12N2O3S
    Formula Weight 216.26
    MDL Number MFCD00005176
    Beilstein 15080
    Merck Index 14,458
    Synonym 6-APA
    Chemical Name or Material 6-Aminopenicillanic acid

    Specifications

    Formula Weight 216.26
    Quantity 10g
    Optical Rotation +275° (c=1 in 0.1N HCl)
    Molecular Formula C8H12N2O3S
    Merck Index 14,458
    Recommended Storage 2° to 8°C
    TSCA TSCA
    Melting Point 200°C (dec.)
    MDL Number MFCD00005176
    CAS 551-16-6
    Beilstein 15080
    Synonym 6-APA
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H334-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P284a-P261-P280-P342+P311-P363-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Wash contaminated clothing before reuse.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number XH8225000
    Chemical Name or Material 6-Aminopenicillanic acid
    Assay 98%

    Description

    Applications
    6-Aminopenicillanic acid is the basic unit in penicillin’s, which is obtained from the fermentation brew of the Penicillium mold. It acts as a starting material for the preparation of many semisynthetic penicillins. Further, it serves as apharmaceutical intermediate. In addition to this, it is used in antibiotics research and experimental application.

    Solubility
    Soluble in water and hydrochoric acid.

    Notes
    Store in a cool place. Incompatible with strong oxidizing agents.

    GHS H Statement
    H334-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause an allergic skin reaction.

    2° to 8°C

    GHS P Statement
    P284a-P261-P280-P342+P311-P363-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Wash contaminated clothing before reuse.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit