Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene, 97%

Manufacturer:  Alfa Aesar™ H3043609

Quantity 10g

 View more versions of this product

Catalog No. AAH3043609


  Check Availability
Add to cart

Specifications

Description

Applications
6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene is used in Synthesis and evaluation of synthetic retinoid derivatives as inducers of stem cell differentiation.

Solubility
Insoluble in water

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

27452-17-1
267.21
6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene
C14H19Br
MFCD05664407

Specifications

27452-17-1
267.21
34° to 37°C
6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene
C14H19Br
MFCD05664407
10g
97%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit