Alfa Aesar™ 6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene, 97%

Manufacturer: Alfa Aesar™ H3043603

Quantity 1g

 View more versions of this product

Catalog No. AAH3043603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 27452-17-1
    Molecular Formula C14H19Br
    Formula Weight 267.21
    MDL Number MFCD05664407
    Chemical Name or Material 6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene

    Specifications

    Formula Weight 267.21
    Molecular Formula C14H19Br
    MDL Number MFCD05664407
    CAS 27452-17-1
    Melting Point 34° to 37°C
    Quantity 1g
    Chemical Name or Material 6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene
    Assay 97%

    Description

    Applications
    6-Bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene is used in Synthesis and evaluation of synthetic retinoid derivatives as inducers of stem cell differentiation.

    Solubility
    Insoluble in water

    Notes
    Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit