Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromo-[1,2,4]triazolo[1,5-a]pyridine, 96%, Alfa Aesar™

Click to view available options
Quantity:
250mg
1g
356560-80-0
C6H4BrN3
198.023
MFCD08689534
CXRXKDSDRWLKTK-UHFFFAOYSA-N
6-bromo-1,2,4 triazolo 1,5-a pyridine,6-bromo 1,2,4 triazolo 1,5-a pyridine,1,2,4 triazolo 1,5-a pyridine, 6-bromo,6-bromo-1,2,4-triazolo-1,5-a-pyridine,6-bromo-1,2,4-triazolo 1,5-a pyridine,1,2,4 triazolo 1,5-a pyridine,6-bromo,6-bromanyl-1,2,4 triazolo 1,5-a pyridine,6-bromo 1,2,4 triazolo 1,5-alpha pyridine,6-bromo-1,2,4 triazolo 1,5-a pyridine,tert-butyl 6e-7-4-4-fluorophenyl-6-isopropyl-2-methyl methylsulfonyl amino pyrimidin-5-yl-3r,5s-3,5-dihydroxyhept-6-enoate
11513934
6-bromo-[1,2,4]triazolo[1,5-a]pyridine
C1=CC2=NC=NN2C=C1Br
356560-80-0
C6H4BrN3
198.023
MFCD08689534
CXRXKDSDRWLKTK-UHFFFAOYSA-N
6-bromo-1,2,4 triazolo 1,5-a pyridine,6-bromo 1,2,4 triazolo 1,5-a pyridine,1,2,4 triazolo 1,5-a pyridine, 6-bromo,6-bromo-1,2,4-triazolo-1,5-a-pyridine,6-bromo-1,2,4-triazolo 1,5-a pyridine,1,2,4 triazolo 1,5-a pyridine,6-bromo,6-bromanyl-1,2,4 triazolo 1,5-a pyridine,6-bromo 1,2,4 triazolo 1,5-alpha pyridine,6-bromo-1,2,4 triazolo 1,5-a pyridine,tert-butyl 6e-7-4-4-fluorophenyl-6-isopropyl-2-methyl methylsulfonyl amino pyrimidin-5-yl-3r,5s-3,5-dihydroxyhept-6-enoate
11513934
6-bromo-[1,2,4]triazolo[1,5-a]pyridine
C1=CC2=NC=NN2C=C1Br
Quantity 1g
Beilstein 10061274
Formula Weight 198.02
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 6-Bromo-[1,2,4]triazolo[1,5-a]pyridine