Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-bromo-1,3-benzoxazol-2(3h)-one, 97%, Thermo Scientific™

Catalog Number p-3880686
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
19932-85-5
C7H4BrNO2
214.02
MFCD00694727
DDNKJFBQMQOIKI-UHFFFAOYSA-N
6-bromobenzo d oxazol-2 3h-one, 6-bromo-2-benzoxazolinone, 6-bromo-1,3-benzoxazol-2 3h-one, 2-benzoxazolinone, 6-bromo, 6-bromobenzoxazolinone, 2 3h-benzoxazolone, 6-bromo, 6-bromo-2 3h-benzoxazolone, cincreasin, 6-bromo-2,3-dihydro-1,3-benzoxazol-2-one, 6-bromobenzoxazol-2 3h-one
29859
CHEBI:77861
6-bromo-3H-1,3-benzoxazol-2-one
BrC1=CC=C2NC(=O)OC2=C1
19932-85-5
C7H4BrNO2
214.02
MFCD00694727
DDNKJFBQMQOIKI-UHFFFAOYSA-N
6-bromobenzo d oxazol-2 3h-one, 6-bromo-2-benzoxazolinone, 6-bromo-1,3-benzoxazol-2 3h-one, 2-benzoxazolinone, 6-bromo, 6-bromobenzoxazolinone, 2 3h-benzoxazolone, 6-bromo, 6-bromo-2 3h-benzoxazolone, cincreasin, 6-bromo-2,3-dihydro-1,3-benzoxazol-2-one, 6-bromobenzoxazol-2 3h-one
29859
CHEBI:77861
6-bromo-3H-1,3-benzoxazol-2-one
BrC1=CC=C2NC(=O)OC2=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 214.02
Percent Purity 97%
Chemical Name or Material 6-Bromo-1, 3-benzoxazol-2(3h)-one
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit