Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromo[1,3]oxazolo[4,5-b]pyridin-2(3H)-one, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
21594-52-5
C6H3BrN2O2
215.01
MFCD08689590
VQPBRWIFFBIRRP-UHFFFAOYSA-N
6-bromo-3h-oxazolo 4,5-b pyridin-2-one,6-bromooxazolo 4,5-b pyridin-2 3h-one,6-bromo-2h,3h-1,3 oxazolo 4,5-b pyridin-2-one,oxazolo 4,5-b pyridin-2 3h-one, 6-bromo,6-bromo 1,3 oxazolo 4,5-b pyridin-2 3h-one,6-bromo-oxazolo 4,5-b pyridin-2 3h-one,6-bromo-3h-1,3 oxazolo 4,5-b pyridin-2-one,pubchem17712,acmc-1cakb,6-bromooxooxazolo 4,5-b pyridine
13614458
6-bromo-3H-[1,3]oxazolo[4,5-b]pyridin-2-one
C1=C2C(=NC=C1Br)NC(=O)O2
21594-52-5
C6H3BrN2O2
215.01
MFCD08689590
VQPBRWIFFBIRRP-UHFFFAOYSA-N
6-bromo-3h-oxazolo 4,5-b pyridin-2-one,6-bromooxazolo 4,5-b pyridin-2 3h-one,6-bromo-2h,3h-1,3 oxazolo 4,5-b pyridin-2-one,oxazolo 4,5-b pyridin-2 3h-one, 6-bromo,6-bromo 1,3 oxazolo 4,5-b pyridin-2 3h-one,6-bromo-oxazolo 4,5-b pyridin-2 3h-one,6-bromo-3h-1,3 oxazolo 4,5-b pyridin-2-one,pubchem17712,acmc-1cakb,6-bromooxooxazolo 4,5-b pyridine
13614458
6-bromo-3H-[1,3]oxazolo[4,5-b]pyridin-2-one
C1=C2C(=NC=C1Br)NC(=O)O2
Packaging Glass bottle
Melting Point 230°C to 232°C
Color Colorless to Amber
Infrared Spectrum Authentic
Quantity 1g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 215.01
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 6-Bromo[1, 3]oxazolo[4, 5-b]pyridin-2(3H)-one
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning