Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromo-1,4-benzodioxane, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
52287-51-1
C8H7BrO2
215.05
LFCURAJBHDNUNG-UHFFFAOYSA-N
6-bromo-1,4-benzodioxane,6-bromo-2,3-dihydrobenzo b 1,4 dioxine,3,4-ethylenedioxybromobenzene,6-bromo-2,3-dihydro-1,4-benzodioxin,1,4-benzodioxin, 6-bromo-2,3-dihydro,4-bromo-1,2-ethylenedioxybenzene,3,4-ethylendioxybromobenzene,6-bromo-2,3-dihydro-benzo 1,4 dioxine,pubchem3736,acmc-209kyk
104141
6-bromo-2,3-dihydro-1,4-benzodioxine
C1COC2=C(O1)C=CC(=C2)Br
52287-51-1
C8H7BrO2
215.05
LFCURAJBHDNUNG-UHFFFAOYSA-N
6-bromo-1,4-benzodioxane,6-bromo-2,3-dihydrobenzo b 1,4 dioxine,3,4-ethylenedioxybromobenzene,6-bromo-2,3-dihydro-1,4-benzodioxin,1,4-benzodioxin, 6-bromo-2,3-dihydro,4-bromo-1,2-ethylenedioxybenzene,3,4-ethylendioxybromobenzene,6-bromo-2,3-dihydro-benzo 1,4 dioxine,pubchem3736,acmc-209kyk
104141
6-bromo-2,3-dihydro-1,4-benzodioxine
C1COC2=C(O1)C=CC(=C2)Br
Boiling Point 259°C to 260°C
Packaging Glass Bottle
Flash Point >110°C
Refractive Index 1.588
Specific Gravity 1.598
Density 1.5980g/mL
Color Colorless to Yellow
Infrared Spectrum Authentic
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 215.05
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 6-Bromo-1, 4-benzodioxane
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 257-817-2