Alfa Aesar™ 6-Bromo-1,4-benzodioxane, 98%

Manufacturer: Alfa Aesar™ B2562503

Quantity 1g

 View more versions of this product

Catalog No. AAB2562503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 52287-51-1
    Molecular Formula C8H7BrO2
    Formula Weight 215.05
    MDL Number MFCD00040750
    Beilstein 6223
    Synonym 4-Bromo-1,2-(ethylenedioxy)benzene; 3,4-(Ethylenedioxy)bromobenzene
    Chemical Name or Material 6-Bromo-1,4-benzodioxane

    Specifications

    Formula Weight 215.05
    Quantity 1g
    Boiling Point 259° to 260°C
    Flash Point >110°C (230°F)
    Molecular Formula C8H7BrO2
    MDL Number MFCD00040750
    Synonym 4-Bromo-1,2-(ethylenedioxy)benzene; 3,4-(Ethylenedioxy)bromobenzene
    CAS 52287-51-1
    Beilstein 6223
    Chemical Name or Material 6-Bromo-1,4-benzodioxane
    Assay 98%

    Description

    Applications
    6-Bromo-1,4-benzodioxane is used as a pharmaceutical intermediate.

    Notes
    Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit