Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Bromo-1-hexanol, 97%

Click to view available options
Quantity:
1g
5g
25g
4286-55-9
C6H13BrO
181.073
MFCD00002983
FCMCSZXRVWDVAW-UHFFFAOYSA-N
6-bromo-1-hexanol,6-bromohexanol,1-hexanol, 6-bromo,6-bromo-hexan-1-ol,hexamethylene bromohydrin,6-bromo-hexanol,6-bromo-l-hexanol,1-bromo-6-hexanol,1-bromohexan-6-ol,6-hydroxyhexyl bromide
77970
6-bromohexan-1-ol
C(CCCBr)CCO
4286-55-9
C6H13BrO
181.073
MFCD00002983
FCMCSZXRVWDVAW-UHFFFAOYSA-N
6-bromo-1-hexanol,6-bromohexanol,1-hexanol, 6-bromo,6-bromo-hexan-1-ol,hexamethylene bromohydrin,6-bromo-hexanol,6-bromo-l-hexanol,1-bromo-6-hexanol,1-bromohexan-6-ol,6-hydroxyhexyl bromide
77970
6-bromohexan-1-ol
C(CCCBr)CCO
Applications
A product for proteomics research. Also used as a reagent in the preparation of nitric oxide-donating analogues of Sulindac (S699215), a non-steroidal anti-inflammatory drug that has the ability to prevent colon cancer.

Solubility
Soluble in chloroform and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Boiling Point 105°C to 106°C (5mmHg)
Flash Point >110°C (230°F)
Quantity 25g
Formula Weight 181.08
Percent Purity 97%
Chemical Name or Material 6-Bromo-1-hexanol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Recommended Storage : 2° to 8°C