Learn More
6-Bromo-1-hexanol, 96%, Thermo Scientific Chemicals
Description
A product for proteomics research. Also used as a reagent in the preparation of nitric oxide-donating analogues of Sulindac (S699215), a non-steroidal anti-inflammatory drug that has the ability to prevent colon cancer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
A product for proteomics research. Also used as a reagent in the preparation of nitric oxide-donating analogues of Sulindac (S699215), a non-steroidal anti-inflammatory drug that has the ability to prevent colon cancer.
Solubility
Soluble in chloroform and methanol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Density | 1.384 |
Boiling Point | 105°C to 106°C (5 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.482 |
Quantity | 1 g |
Solubility Information | Soluble in chloroform and methanol. |
Formula Weight | 181.08 |
Percent Purity | 97% |
Chemical Name or Material | 6-Bromo-1-hexanol |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
TSCA | Yes |
Recommended Storage | Keep cold |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.