Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromo-1-hexene, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
2695-47-8
C6H11Br
163.06
MFCD00000269
RIMXEJYJXDBLIE-UHFFFAOYSA-N
6-bromo-1-hexene,5-hexenyl bromide,1-hexene, 6-bromo,1-bromo-5-hexene,6-bromohexene,6-bromo-hexene,6-bromo-hex-1-ene,l-bromo-5-hexene,6-bromo-1-hexen,1-bromohex-5-ene
75906
6-bromohex-1-ene
C=CCCCCBr
2695-47-8
C6H11Br
163.06
MFCD00000269
RIMXEJYJXDBLIE-UHFFFAOYSA-N
6-bromo-1-hexene,5-hexenyl bromide,1-hexene, 6-bromo,1-bromo-5-hexene,6-bromohexene,6-bromo-hexene,6-bromo-hex-1-ene,l-bromo-5-hexene,6-bromo-1-hexen,1-bromohex-5-ene
75906
6-bromohex-1-ene
C=CCCCCBr
Specific Gravity 1.217
Quantity 5g
Density 1.2170g/mL
Infrared Spectrum Authentic
Refractive Index 1.4645 to 1.4665 (20°C, 589nm)
Color Yellow
Flash Point 52°C
Packaging Glass bottle
Boiling Point 47.0°C to 51.0°C (16.0 mmHg)
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula Br(CH2)4CH=CH2
Formula Weight 163.06
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 6-Bromo-1-hexene
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/

GHS Signal Word: Warning

EINECSNumber : 220-267-9