Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromo-1-methyl-1H-indazole, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
590417-94-0
C8H7BrN2
211.06
SVVSOIGNROPKMS-UHFFFAOYSA-N
6-bromo-1-methyl-1h-indazole,1-methyl-6-bromoindazole,1h-indazole, 6-bromo-1-methyl,pubchem19529,acmc-209m9p,6-bromo-1-methyl-indazole,indazole, 6-bromo-1-methyl
22558675
6-bromo-1-methylindazole
CN1C2=C(C=CC(=C2)Br)C=N1
590417-94-0
C8H7BrN2
211.06
SVVSOIGNROPKMS-UHFFFAOYSA-N
6-bromo-1-methyl-1h-indazole,1-methyl-6-bromoindazole,1h-indazole, 6-bromo-1-methyl,pubchem19529,acmc-209m9p,6-bromo-1-methyl-indazole,indazole, 6-bromo-1-methyl
22558675
6-bromo-1-methylindazole
CN1C2=C(C=CC(=C2)Br)C=N1
Melting Point 48°C to 50°C
Packaging Glass Bottle
Quantity 1g
Assay Percent Range 96% min. (GC)
Formula Weight 211.06
Percent Purity 97%
Chemical Name or Material 6-Bromo-1-methyl-1H-indazole

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning