Alfa Aesar™ 6-Bromo-1-tetralone, 96%

Manufacturer: Alfa Aesar™ H52381MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52381MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 66361-67-9
    Molecular Formula C10H9BrO
    Formula Weight 225.09
    MDL Number MFCD04114378
    Synonym 6-Bromo-3,4-dihydro-2H-naphthalen-1-one
    Chemical Name or Material 6-Bromo-1-tetralone

    Specifications

    Formula Weight 225.09
    Melting Point 40° to 42°C
    Quantity 250mg
    Molecular Formula C10H9BrO
    MDL Number MFCD04114378
    Synonym 6-Bromo-3,4-dihydro-2H-naphthalen-1-one
    Recommended Storage 2° to 8°C
    CAS 66361-67-9
    Chemical Name or Material 6-Bromo-1-tetralone
    Assay 96%

    Description

    Applications
    6-bromo-1-tetralone used in Wittig-Horner reaction.

    Notes
    Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, heat.

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit