Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Bromo-2(3H)-benzoxazolone, 97%

Manufacturer:  Alfa Aesar™ H3398403

Quantity 1g

 View more versions of this product

Catalog No. AAH3398403


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

19932-85-5
214
6-Bromo-2-benzoxazolinone
C7H4BrNO2
MFCD00694727
6-Bromo-2(3H)-benzoxazolone

Specifications

19932-85-5
214
193° to 195°C
1g
97%
C7H4BrNO2
MFCD00694727
6-Bromo-2-benzoxazolinone
6-Bromo-2(3H)-benzoxazolone

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P301+P312-P337+P313-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DM4960000

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit