Alfa Aesar™ 6-Bromo-2-chloroquinoline, 96%

Manufacturer: Alfa Aesar™ H5489506

Quantity 5g

 View more versions of this product

Catalog No. AAH5489506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1810-71-5
    Molecular Formula C9H5BrClN
    Formula Weight 242.5
    MDL Number MFCD04115272
    Chemical Name or Material 6-Bromo-2-chloroquinoline

    Specifications

    Formula Weight 242.5
    Molecular Formula C9H5BrClN
    MDL Number MFCD04115272
    Quantity 5g
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    CAS 1810-71-5
    Melting Point 148° to 150°C
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 6-Bromo-2-chloroquinoline
    Assay 96%

    Description

    Applications
    It is used as a pharmaceutical intermediate.

    Solubility
    Soluble in Chloroform and Methanol

    Notes
    Store at room temperature with proper ventilation.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit