Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Bromo-2-methyl-3-nitropyridine, 97%

Click to view available options
Quantity:
5g
25g
22282-96-8
C6H5BrN2O2
217.022
MFCD03095219
PCNVKBBKROTUNS-UHFFFAOYSA-N
2-bromo-5-nitro-6-picoline,2-bromo-6-methyl-5-nitropyridine,2-bromo-5-nitro-6-methylpyridine,6-bromo-2-methyl-3-nitro-pyridine,6-bromo-3-nitro-2-picoline,6-bromo-3-nitropicoline,6-brom-2-methyl-3-nitro-pyridin,pubchem2968,acmc-209fuh,ksc549a9p
24728014
6-bromo-2-methyl-3-nitropyridine
CC1=C(C=CC(=N1)Br)[N+](=O)[O-]
22282-96-8
C6H5BrN2O2
217.022
MFCD03095219
PCNVKBBKROTUNS-UHFFFAOYSA-N
2-bromo-5-nitro-6-picoline,2-bromo-6-methyl-5-nitropyridine,2-bromo-5-nitro-6-methylpyridine,6-bromo-2-methyl-3-nitro-pyridine,6-bromo-3-nitro-2-picoline,6-bromo-3-nitropicoline,6-brom-2-methyl-3-nitro-pyridin,pubchem2968,acmc-209fuh,ksc549a9p
24728014
6-bromo-2-methyl-3-nitropyridine
CC1=C(C=CC(=N1)Br)[N+](=O)[O-]
Quantity 5g
Melting Point 67°C to 72°C
Beilstein 135841
Formula Weight 217.02
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 6-Bromo-2-methyl-3-nitropyridine

storageNote1 : 2° to 8°C