Alfa Aesar™ 6-Bromo-2-methyl-4H-3,1-benzoxazin-4-one

Manufacturer: Alfa Aesar™ H5058503

Quantity 1g

 View more versions of this product

Catalog No. AAH5058503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 19165-25-4
    Molecular Formula C9H6BrNO2
    Formula Weight 240.06
    MDL Number MFCD00115891
    Chemical Name or Material 6-Bromo-2-methyl-4H-3,1-benzoxazin-4-one

    Specifications

    Formula Weight 240.06
    Molecular Formula C9H6BrNO2
    MDL Number MFCD00115891
    CAS 19165-25-4
    Quantity 1g
    Chemical Name or Material 6-Bromo-2-methyl-4H-3,1-benzoxazin-4-one

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit