Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromo-2-picoline, 97%, Acros Organics

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
5315-25-3
C6H6BrN
172.02
MFCD00040743
SOHDPICLICFSOP-UHFFFAOYSA-N
2-bromo-6-picoline,6-bromo-2-picoline,6-bromo-2-methylpyridine,2-bromo-6-methyl-pyridine,2-bromo-6-methyl pyridine,2-brom-6-methylpyridin,pyridine, 2-bromo-6-methyl,2-bromo-6-methy pyridine,6-bromopicoline,6-methyl-2-bromopyridine
79205
2-bromo-6-methylpyridine
CC1=NC(=CC=C1)Br
5315-25-3
C6H6BrN
172.02
MFCD00040743
SOHDPICLICFSOP-UHFFFAOYSA-N
2-bromo-6-picoline,6-bromo-2-picoline,6-bromo-2-methylpyridine,2-bromo-6-methyl-pyridine,2-bromo-6-methyl pyridine,2-brom-6-methylpyridin,pyridine, 2-bromo-6-methyl,2-bromo-6-methy pyridine,6-bromopicoline,6-methyl-2-bromopyridine
79205
2-bromo-6-methylpyridine
CC1=NC(=CC=C1)Br
Specific Gravity 1.512
Boiling Point 102°C to 103°C (20.0mmHg)
Flash Point 97°C
Refractive Index 1.5620 to 1.564
Density 1.5120g/mL
Color Colorless to Yellow
Infrared Spectrum Authentic
Quantity 10g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 172.02
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 6-Bromo-2-picoline
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 226-173-4