Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromo-2-pyridinecarbonitrile, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
122918-25-6
C6H3BrN2
183.01
MFCD00234142
HNEBPTAKURBYRM-UHFFFAOYSA-N
6-bromopicolinonitrile,6-bromo-2-pyridinecarbonitrile,2-bromo-6-cyanopyridine,6-bromo-2-cyanopyridine,2-pyridinecarbonitrile, 6-bromo,2-cyano-6-bromopyridine,2-bromopyridine-6-carbonitrile,pubchem14367,2-bromo-6-cyanopyridin,2-bromo-6-cyano pyridine
15100859
6-bromopyridine-2-carbonitrile
C1=CC(=NC(=C1)Br)C#N
122918-25-6
C6H3BrN2
183.01
MFCD00234142
HNEBPTAKURBYRM-UHFFFAOYSA-N
6-bromopicolinonitrile,6-bromo-2-pyridinecarbonitrile,2-bromo-6-cyanopyridine,6-bromo-2-cyanopyridine,2-pyridinecarbonitrile, 6-bromo,2-cyano-6-bromopyridine,2-bromopyridine-6-carbonitrile,pubchem14367,2-bromo-6-cyanopyridin,2-bromo-6-cyano pyridine
15100859
6-bromopyridine-2-carbonitrile
C1=CC(=NC(=C1)Br)C#N
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 98.5°C to 101.0°C
Color White
Quantity 1g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 183.01
Physical Form Low Melting Solid or Crystalline Powder
Percent Purity 98%
Chemical Name or Material 6-Bromo-2-pyridinecarbonitrile
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning