Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromo-3-methyl-1,3-benzoxazol-2(3H)-one, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
67927-44-0
C8H6BrNO2
228.05
MFCD00694789
OASANCDKMGODJO-UHFFFAOYSA-N
6-bromo-3-methyl-2,3-dihydro-1,3-benzoxazol-2-one,6-bromo-3-methylbenzo d oxazol-2 3h-one,6-bromo-3-methyl-1,3-benzoxazol-2 3h-one,bromomethyldihydrobenzoxazolone,6-bromo-n-methylbenzoxazolone,6-bromo-3-methyl-2 3h-benzoxazolone,6-bromo-3-methyl-3h-benzooxazol-2-one,6-bromo-3-methyl-1?3-benzoxazol-2 3h-one
379590
6-bromo-3-methyl-1,3-benzoxazol-2-one
CN1C2=C(C=C(C=C2)Br)OC1=O
67927-44-0
C8H6BrNO2
228.05
MFCD00694789
OASANCDKMGODJO-UHFFFAOYSA-N
6-bromo-3-methyl-2,3-dihydro-1,3-benzoxazol-2-one,6-bromo-3-methylbenzo d oxazol-2 3h-one,6-bromo-3-methyl-1,3-benzoxazol-2 3h-one,bromomethyldihydrobenzoxazolone,6-bromo-n-methylbenzoxazolone,6-bromo-3-methyl-2 3h-benzoxazolone,6-bromo-3-methyl-3h-benzooxazol-2-one,6-bromo-3-methyl-1?3-benzoxazol-2 3h-one
379590
6-bromo-3-methyl-1,3-benzoxazol-2-one
CN1C2=C(C=C(C=C2)Br)OC1=O
Melting Point 149°C to 150°C
Packaging Glass bottle
Quantity 1g
Infrared Spectrum Authentic
Color White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 228.05
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 6-Bromo-3-methyl-1, 3-benzoxazol-2(3H)-one
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning