Learn More
Medchemexpress LLC 6-Bromo-4-chloroquinoline | 65340-70-7 | 99.96% | C9H5BrClN | 10 G
SDP
Supplier:  Medchemexpress LLC HY7514410G
Encompass_Preferred
This compound is a drug intermediate used for synthesizing various active compounds. It is classified as an acute oral toxicant (Category 4), causes skin corrosion/irritation (Category 2), serious eye damage/eye irritation (Category 2A), and specific target organ toxicity - single exposure; respiratory tract irritation (Category 3). It is stable under recommended storage conditions, appearing as a solid with a melting point between 110-112 °C. It should be handled by suitably qualified and experienced scientists in appropriately equipped and authorized facilities for research use only.
  • Appears as a solid
  • White to light yellow color
  • Molecular weight of 242.50
  • Stable under recommended storage conditions
  • Suitable for synthesizing various active compounds
  • Melting/freezing point between 110-112 °C
Catalog No. 50-004-34625