Alfa Aesar™ 6-Bromo-4-methyl-3-nitroquinoline, 96%

Manufacturer: Alfa Aesar™ H5476903

Quantity 1g

 View more versions of this product

Catalog No. AAH5476903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1185310-49-9
    Molecular Formula C10H7BrN2O2
    Formula Weight 267.08
    MDL Number MFCD12761249
    Beilstein 19927450
    Chemical Name or Material 6-Bromo-4-methyl-3-nitroquinoline

    Specifications

    Formula Weight 267.08
    Molecular Formula C10H7BrN2O2
    MDL Number MFCD12761249
    CAS 1185310-49-9
    Melting Point 152° to 155°C
    Quantity 1g
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Recommended Storage 2° to 8°C
    Beilstein 19927450
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 6-Bromo-4-methyl-3-nitroquinoline
    Assay 96%

    Description

    Applications
    6-Bromo-4-methyl-3-nitroquinoline is used as pharmaceutical intermediate.

    Notes
    Air sensitive. Incompatible with oxidizing agents and heat.

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit