Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromo-8-fluoroquinoline, 97%, Thermo Scientific™

Click to view available options
Quantity:
1g
5g
220513-46-2
C9H5BrFN
226.05
DDCOYWOYIUEOHQ-UHFFFAOYSA-N
acmc-1crnu, 6-bromo-8-fluoro-quinoline, 6-bromo-8-fluoroquinoline, 6-bromo-8-fluoro-1-azanaphthalene
46738885
6-bromo-8-fluoroquinoline
C1=CC2=CC(=CC(=C2N=C1)F)Br
220513-46-2
C9H5BrFN
226.05
DDCOYWOYIUEOHQ-UHFFFAOYSA-N
acmc-1crnu, 6-bromo-8-fluoro-quinoline, 6-bromo-8-fluoroquinoline, 6-bromo-8-fluoro-1-azanaphthalene
46738885
6-bromo-8-fluoroquinoline
C1=CC2=CC(=CC(=C2N=C1)F)Br
Melting Point 103°C to 106°C
Assay Percent Range 96% min. (GC)
Quantity 5g
Formula Weight 226.05
Percent Purity 97%
Chemical Name or Material 6-Bromo-8-fluoroquinoline

GHS Signal Word: Warning

RUO – Research Use Only