Alfa Aesar™ 6-Bromohexanoyl chloride, 97%

Manufacturer: Alfa Aesar™ A1106336

Quantity 500g

 View more versions of this product

Catalog No. AAA1106336

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 22809-37-6
    Molecular Formula C6H10BrClO
    Linear Formula Br(CH2)5COCl
    Formula Weight 213.5
    MDL Number MFCD00000761
    UN Number UN3265
    Beilstein 1749739
    Synonym 6-Bromocaproyl chloride
    Chemical Name or Material 6-Bromohexanoyl chloride

    Specifications

    Formula Weight 213.5
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C6H10BrClO
    CAS 22809-37-6
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    TSCA TSCA
    MDL Number MFCD00000761
    UN Number UN3265
    Beilstein 1749739
    Odor Acrid
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Linear Formula Br(CH2)5COCl
    Synonym 6-Bromocaproyl chloride
    Boiling Point 119° to 120°C (9mmHg)
    Quantity 500g
    Flash Point >110°C (230°F)
    Chemical Name or Material 6-Bromohexanoyl chloride
    Assay 97%

    Description

    Applications
    6-Bromohexanoyl chloride is used as intermediates, organic synthesis, chemical research, agrochemicals and dyestuff.

    Solubility
    Reacts with water.

    Notes
    Store in cool, dry conditions in well sealed containers. Keep container tightly closed. Moisture Sensitive

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit