Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromoimidazo[1,2-a]pyridine-3-carboxylic acid, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
944896-42-8
C8H5BrN2O2
241.04
SDKJLYHPCBMDAD-UHFFFAOYSA-N
6-bromoimidazo 1,2-a pyridine-3-carboxylic acid,6-bromoimidazo 1,2-a pyridine-3-carboxylicacid,6-bromoh-imidazo 1,2-a pyridine-3-carboxylic acid,6-bromo-1h-imidazo 1,2-a pyridine-3-carboxylic acid,6-bromo-3-carboxyimidazo 1,2-a pyridine,6-bromo-imidazo 1,2-a pyridine-3-carboxylic acid,imidazo 1,2-a pyridine-3-carboxylic acid, 6-bromo
22031912
6-bromoimidazo[1,2-a]pyridine-3-carboxylic acid
C1=CC2=NC=C(N2C=C1Br)C(=O)O
944896-42-8
C8H5BrN2O2
241.04
SDKJLYHPCBMDAD-UHFFFAOYSA-N
6-bromoimidazo 1,2-a pyridine-3-carboxylic acid,6-bromoimidazo 1,2-a pyridine-3-carboxylicacid,6-bromoh-imidazo 1,2-a pyridine-3-carboxylic acid,6-bromo-1h-imidazo 1,2-a pyridine-3-carboxylic acid,6-bromo-3-carboxyimidazo 1,2-a pyridine,6-bromo-imidazo 1,2-a pyridine-3-carboxylic acid,imidazo 1,2-a pyridine-3-carboxylic acid, 6-bromo
22031912
6-bromoimidazo[1,2-a]pyridine-3-carboxylic acid
C1=CC2=NC=C(N2C=C1Br)C(=O)O
Packaging Glass Bottle
Quantity 1g
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Formula Weight 241.04
Percent Purity 97%
Chemical Name or Material 6-Bromoimidazo[1,2-a]pyridine-3-carboxylic acid

GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning