Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Bromoindirubin-3'-oxime, 97%

Click to view available options
Quantity:
10mg
50mg
667463-62-9
C16H10BrN3O2
356.179
MFCD08705318
WNWSUJQVZJJGLF-UHFFFAOYSA-N
bio,6-bromo-3-3-hydroxyamino indol-2-ylidene-1h-indol-2-one,z,3e-6'-bromo-3-hydroxyimino-1h,1'h-2,3'-biindolyliden-2'-one,2h-indol-2-one, 6-bromo-3-3e-1,3-dihydro-3-hydroxyimino-2h-indol-2-ylidene-1,3-dihydro-, 3z,6-bromo-3-3-hydroxyamino indol-2-ylidene indolin-2-one,2z,3e-6'-bromo-3-hydroxyimino-2,3'-biindolinylidene-2'-one,6-bromo-3-3-hydroxyamino-2h-indol-2-ylidene-1,3-dihydro-2h-indol-2-one
24906282
6-bromo-3-[3-(hydroxyamino)indol-2-ylidene]-1H-indol-2-one
C1=CC2=C(C(=C3C4=C(C=C(C=C4)Br)NC3=O)N=C2C=C1)NO
667463-62-9
C16H10BrN3O2
356.179
MFCD08705318
WNWSUJQVZJJGLF-UHFFFAOYSA-N
bio,6-bromo-3-3-hydroxyamino indol-2-ylidene-1h-indol-2-one,z,3e-6'-bromo-3-hydroxyimino-1h,1'h-2,3'-biindolyliden-2'-one,2h-indol-2-one, 6-bromo-3-3e-1,3-dihydro-3-hydroxyimino-2h-indol-2-ylidene-1,3-dihydro-, 3z,6-bromo-3-3-hydroxyamino indol-2-ylidene indolin-2-one,2z,3e-6'-bromo-3-hydroxyimino-2,3'-biindolinylidene-2'-one,6-bromo-3-3-hydroxyamino-2h-indol-2-ylidene-1,3-dihydro-2h-indol-2-one
24906282
6-bromo-3-[3-(hydroxyamino)indol-2-ylidene]-1H-indol-2-one
C1=CC2=C(C(=C3C4=C(C=C(C=C4)Br)NC3=O)N=C2C=C1)NO
Quantity 50mg
Color Red
Sensitivity Air & Light Sensitive
Formula Weight 356.17
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 6-Bromoindirubin-3'-oxime

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : NM3241900

storageNote1 : 2° to 8°C