Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Bromoindole-3-carboxaldehyde, 95%

Click to view available options
Quantity:
1g
5g
17826-04-9
C9H6BrNO
224.057
MFCD00792689
WCCLQCBKBPTODV-UHFFFAOYSA-N
6-bromoindole-3-carboxaldehyde,6-bromoindole-3-carbaldehyde,6-bromo-3-formylindole,6-bromo-1h-indole-3-carboxaldehyde,6-bromindol-3-carbaldehyde,1h-indole-3-carboxaldehyde, 6-bromo,pubchem7691,acmc-209ee9,ksc536o3d,wcclqcbkbptodv-uhfffaoysa
2794830
6-bromo-1H-indole-3-carbaldehyde
C1=CC2=C(C=C1Br)NC=C2C=O
17826-04-9
C9H6BrNO
224.057
MFCD00792689
WCCLQCBKBPTODV-UHFFFAOYSA-N
6-bromoindole-3-carboxaldehyde,6-bromoindole-3-carbaldehyde,6-bromo-3-formylindole,6-bromo-1h-indole-3-carboxaldehyde,6-bromindol-3-carbaldehyde,1h-indole-3-carboxaldehyde, 6-bromo,pubchem7691,acmc-209ee9,ksc536o3d,wcclqcbkbptodv-uhfffaoysa
2794830
6-bromo-1H-indole-3-carbaldehyde
C1=CC2=C(C=C1Br)NC=C2C=O
Quantity 1g
Melting Point 202°C to 206°C
Beilstein 1526171
Formula Weight 224.06
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 6-Bromoindole-3-carboxaldehyde

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning