Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromoisatin, 95%, Acros Organics™

Click to view available options
Quantity:
5g
25g
6326-79-0
C8H4BrNO2
226.03
HVPQMLZLINVIHW-UHFFFAOYSA-N
6-bromoisatin,6-bromoindoline-2,3-dione,6-bromo-isatin,1h-indole-2,3-dione, 6-bromo,6-bromoindole-2,3-dione,6-bromo-2,3-dihydro-1h-indole-2,3-dione,6-bromo-1h-benzo d azolidine-2,3-dione,6-bromoindolin-2,3-dione,6-bromo-2,3-indolinedione
95716
6-bromo-1H-indole-2,3-dione
C1=CC2=C(C=C1Br)NC(=O)C2=O
6326-79-0
C8H4BrNO2
226.03
HVPQMLZLINVIHW-UHFFFAOYSA-N
6-bromoisatin,6-bromoindoline-2,3-dione,6-bromo-isatin,1h-indole-2,3-dione, 6-bromo,6-bromoindole-2,3-dione,6-bromo-2,3-dihydro-1h-indole-2,3-dione,6-bromo-1h-benzo d azolidine-2,3-dione,6-bromoindolin-2,3-dione,6-bromo-2,3-indolinedione
95716
6-bromo-1H-indole-2,3-dione
C1=CC2=C(C=C1Br)NC(=O)C2=O
Quantity 25g
Infrared Spectrum Authentic
Melting Point 274.0°C
Assay Percent Range 94% min. (HPLC)
Formula Weight 226.03
Percent Purity 95%
Chemical Name or Material 6-Bromoisatin

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause an allergic skin reaction.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a

GHS Signal Word: Warning