Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromonicotinic acid, 96%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
6311-35-9
C6H4BrNO2
202.01
MFCD01927100
JDJBRMNTXORYEN-UHFFFAOYSA-N
6-bromonicotinic acid,6-bromo nicotinic acid,2-bromo-5-pyridinecarboxylic acid,2-bromopyridine-5-carboxylic acid,6-bromo-3-pyridine carboxylic acid,6-bromonicotinicacid,6-bromo-nicotinic acid,6-bromo-3-pyridinecarboxylic acid,2-bromo-5-pyridine carboxylic acid,3-pyridinecarboxylic acid, 6-bromo
238932
6-bromopyridine-3-carboxylic acid
C1=CC(=NC=C1C(=O)O)Br
6311-35-9
C6H4BrNO2
202.01
MFCD01927100
JDJBRMNTXORYEN-UHFFFAOYSA-N
6-bromonicotinic acid,6-bromo nicotinic acid,2-bromo-5-pyridinecarboxylic acid,2-bromopyridine-5-carboxylic acid,6-bromo-3-pyridine carboxylic acid,6-bromonicotinicacid,6-bromo-nicotinic acid,6-bromo-3-pyridinecarboxylic acid,2-bromo-5-pyridine carboxylic acid,3-pyridinecarboxylic acid, 6-bromo
238932
6-bromopyridine-3-carboxylic acid
C1=CC(=NC=C1C(=O)O)Br
Melting Point 179°C to 181°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1g
Color White
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 202.01
Physical Form Crystalline Powder
Percent Purity 96%
Chemical Name or Material 6-Bromonicotinic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES.

GHS Signal Word: Warning