Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 6-Bromophthalazin-1(4H)-one, 98%

Click to view available options
Quantity:
250mg
1g
5g
75884-70-7
C8H5BrN2O
225.045
MFCD09264003
QMONLZVJOOMKRW-UHFFFAOYSA-N
6-bromophthalazin-1 2h-one,6-bromophthalazin-1-ol,6-bromo-1,2-dihydrophthalazin-1-one,1 2h-phthalazinone,6-bromo,1 2h-phthalazinone, 6-bromo,ksc642c5d,6-bromophthalazine-1 2h-one,6-bromanyl-2h-phthalazin-1-one,6-bromophthalazine-1 2h-one
11535918
6-bromo-2H-phthalazin-1-one
C1=CC2=C(C=C1Br)C=NNC2=O
75884-70-7
C8H5BrN2O
225.045
MFCD09264003
QMONLZVJOOMKRW-UHFFFAOYSA-N
6-bromophthalazin-1 2h-one,6-bromophthalazin-1-ol,6-bromo-1,2-dihydrophthalazin-1-one,1 2h-phthalazinone,6-bromo,1 2h-phthalazinone, 6-bromo,ksc642c5d,6-bromophthalazine-1 2h-one,6-bromanyl-2h-phthalazin-1-one,6-bromophthalazine-1 2h-one
11535918
6-bromo-2H-phthalazin-1-one
C1=CC2=C(C=C1Br)C=NNC2=O
Quantity 250mg
Beilstein 10426132
Formula Weight 225.04
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 6-Bromophthalazin-1(4H)-one

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning