Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromopicolinic acid, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
21190-87-4
C6H4BrNO2
202.01
XURXQNUIGWHWHU-UHFFFAOYSA-N
6-bromopicolinic acid,6-bromo-2-pyridinecarboxylic acid,6-bromo-2-pyridine carboxylic acid,2-pyridinecarboxylic acid, 6-bromo,2-bromo pyridine-6-carboxylic acid,2-bromopyridine-6-carboxylic acid,2-bromo-6-pyridinecarboxylic acid,6-bromo-pyridine-2-carboxylic acid,6-bromopicolinicacid,pubchem2422
593919
6-bromopyridine-2-carboxylic acid
C1=CC(=NC(=C1)Br)C(=O)O
21190-87-4
C6H4BrNO2
202.01
XURXQNUIGWHWHU-UHFFFAOYSA-N
6-bromopicolinic acid,6-bromo-2-pyridinecarboxylic acid,6-bromo-2-pyridine carboxylic acid,2-pyridinecarboxylic acid, 6-bromo,2-bromo pyridine-6-carboxylic acid,2-bromopyridine-6-carboxylic acid,2-bromo-6-pyridinecarboxylic acid,6-bromo-pyridine-2-carboxylic acid,6-bromopicolinicacid,pubchem2422
593919
6-bromopyridine-2-carboxylic acid
C1=CC(=NC(=C1)Br)C(=O)O
Infrared Spectrum Authentic
Color Tan to White
Packaging Glass bottle
Melting Point 192.0°C to 194.0°C
Quantity 1g
Assay Percent Range 98%
Formula Weight 202.01
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 6-Bromopicolinic acid